Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, Tool Steel, 14.28 mm, 2.0000 in. OAL, 1 ea
Supplier: MSC 68105337
This Class ZZ Plus plug gage is designed for precision inspection and measurement applications. Crafted from durable tool steel, it serves as a reliable tool for Go inspection, setting micrometers, and verifying verniers and other measuring devices. The gage ensures consistent results in metalworking and quality control environments.
- Diameter of 14.28 mm
- Overall length of 2.0000 in.
- Constructed from tool steel for enhanced durability
- Class ZZ tolerance with a plus direction
- Tolerance of 0.00508 mm
- Reversible style for extended service life
- Surface finish of 10 micro
- Includes certificate of accuracy with NIST traceability
- Manufactured to ASME B89.1 standards