Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 13.36 mm Diameter, Tool Steel, 2.0000 in. Length, 1 ea
Supplier:  MSC 68104413
Encompass
The Class ZZ Plus plug gage is designed for precision measurements, including go inspection and the setting of micrometers, verniers, and other measuring devices. This gage is constructed from tool steel and features a 13.36 mm diameter with a 0.00508 mm tolerance to ensure measurement accuracy in industrial applications.
  • Manufactured from tool steel for durability
  • Overall length of 2.0000 in.
  • Includes a certificate of accuracy with NIST traceability
  • Compliant with ASME B89.1 standards
  • Surface finish of 10 micro
Catalog No. 50-005-11146