Learn More
MSC Vermont Gage Pin Gage, 22.00 mm, Class ZZ Minus, Steel, 1 ea
Supplier:  MSC 40449456
Encompass
This Class ZZ minus plug and pin gage is a high-precision measurement tool designed for checking hole sizes, setting micrometers, and calibrating various measuring devices. Constructed from steel, it features a 22.00 mm diameter and a 2 in. overall length.
  • Manufactured for use in NoGo inspection applications
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 microinch surface finish
  • Hardened steel construction with 60 to 65 Rc hardness
  • Reversible style member for extended tool life
Catalog No. 50-005-16657