Learn More
MSC Vermont Gage Plug and Pin Gage, Class ZZ, Minus, 12.39 mm Diameter, 2 in. Length, 1 ea
Supplier:  MSC 40439846
Encompass
This Class ZZ Minus plug and pin gage is a high-precision measurement tool designed for NoGo inspection, setting micrometers, and calibrating verniers and other measuring devices. It provides reliable accuracy with a 12.39 mm diameter and an overall length of 2 in.
  • Class ZZ minus tolerance of 0.00508 mm
  • Constructed from durable tool steel
  • Hardness rating of 60 to 65 Rc
  • Finish of 10 microinch or better
  • Roundness within 0.0025 mm
  • Includes certificate of accuracy with NIST traceability
  • Marked with size and direction of tolerance
Catalog No. 50-005-16537