Learn More
MSC Vermont Gage Class ZZ Pin Gage, 6.76 mm, 2 in., 1 ea
Supplier:  MSC 40434219
Encompass
This Class ZZ plug and pin gage is engineered for precise measurement and inspection tasks, including NoGo inspection and the setting of micrometers and verniers. The 6.76 mm diameter tool is constructed from 52100 tool steel, ensuring durability and consistent performance in industrial applications.
  • Features 2 in. of usable gaging surface for checking hole depths
  • Reversible design allows for extended gage life
  • Heat-treated to 60 to 62 Rc for superior wear resistance
  • Centerless lapped to a 10 microinch or better surface finish
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-13969