Learn More
MSC Vermont Gage Class ZZ Pin Gage, 52100 Tool Steel, 4.66 mm Diameter, 2.0000 in. Length, 1 ea.
Supplier:  MSC 68095710
Encompass
The Class ZZ Plus Pin Gage is a precision measuring tool designed for accurate Go inspection, calibration of micrometers, and verifying other measuring devices. This pin gage is manufactured from durable 52100 tool steel and heat-treated for long-lasting performance in industrial measurement applications.
  • Constructed from 52100 tool steel
  • Hardness rated at 60 to 65 Rc
  • Finished to 10 micro or better
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Overall length of 2.0000 in.
Catalog No. 50-005-12843