Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 16.68 mm Diameter, 2.0000 in. Overall Length, 1 ea
Supplier:  MSC 68107739
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications, including go inspections, setting micrometers, and verifying verniers or other measuring instruments. Manufactured from durable tool steel, it offers reliable performance for rigorous inspection tasks.
  • Diameter of 16.68 mm
  • Overall length of 2.0000 in.
  • Tolerance of 0.00508 mm
  • Includes certificate of accuracy with NIST traceability
  • Constructed from tool steel for durability
  • Coating or finish of 10 micro
  • Conforms to ASME B89.1 standards
Catalog No. 50-005-12771