Learn More
MSC Vermont Gage Class ZZ Plus Pin Gage, Tool Steel, 4.79 mm Diameter, 2.0000 in. OAL, 1 ea.
Supplier:  MSC 68095843
Encompass
The Class ZZ Plus plug gage is a precision measurement tool with a diameter of 4.79 mm and an overall length of 5.08 cm (2.0000 in.). Designed for reliable go inspection, this tool steel gage is suitable for setting micrometers, verniers, and various other measuring devices. It is manufactured to meet rigorous quality standards and includes a certificate of accuracy with NIST traceability to ensure reliable measurement performance.
  • Tool steel construction provides durability with a hardness of 60 to 65 Rc.
  • Reversible style offers flexibility in application.
  • Calibration accredited certificate of accuracy included with each gage.
  • Finished to 10 microinch for high-precision surface quality.
  • System of measurement is metric with a tolerance of 0.00508 mm.
  • Made in the U.S.A.
Catalog No. 50-005-12642