Learn More
TARGETMOL CHEMICALS INC VT103 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T620775MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VT103 is an orally active and selective inhibitor of TEAD1 protein palmitoylation and is an analog of VT101. VT103 has potential antitumor activity by inhibiting YAP/TAZ-TEAD-promoted gene transcriptionment blocking TEAD auto-palmitoylation and blocking the interaction between YAP/TAZ and TEAD.VT103 can be used in the study of HER2-positive breast cancer prostate cancer and triple-negative breast cancer. purity: 98%
Catalog No. 50-257-9486