Learn More
Selleck Chemical LLC VT104 E4655-100MG
SDP
Supplier:  Selleck Chemical LLC E4655100MG
Encompass_Preferred
VT104 is an inhibitor of TEAD auto-palmitoylation that effectively blocks the palmitoylation of TEAD1 and TEAD3 proteins It selectively inhibits YAP/TAZ-TEAD-promoted gene transcription and exhibits strong antitumor and anti-proliferative effects
Catalog No. 50-000-38960