Learn More
Medchemexpress LLC HY-134957 5mg , VT107 CAS:2417718-63-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1349575MG
Encompass_Preferred
HY-134957 5mg VT107 CAS: 2417718-63-7 VT-107, as an analogous to VT104, is an orally active and potent pan-TEAD auto-palmitoylation inhibitor. VT-107 can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9012