Learn More
Cayman Chemical VTP27999trIfluoroacetAT s 5mg
SDP
Supplier:  Cayman Chemical 276895 MG
Encompass_Preferred
A renin inhibitor (IC50 = 0.47 nM for the recombinant human enzyme); selective for renin over β-secretase, cathepsin D, and cathepsin E at 10 µM; >1,000-fold selective for renin over a panel of >250 receptors, ion channels, and enzymes; reduces mean arterial pressure in a double-transgenic rat model of angiotensin II-dependent hypertension at 10 mg/kg
Catalog No. 50-294-0834