Learn More
Medchemexpress LLC HY-114162 5mg Medchemexpress, VTP50469 CAS:2169916-18-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1141625MG
Encompass_Preferred
Medchemexpress, HY-114162 5mg VTP50469 CAS:2169916-18-9 VTP50469 is a potent, highly selective and orally active Menin-MLL interaction inhibitor with a Ki of 104 pM. VTP50469 has potently anti-leukemia activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8832