Learn More
Medchemexpress LLC Vu 0238429 | 1160247-92-6 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1215710MM 1ML
Encompass_Preferred
Vu 0238429 is a positive allosteric modulator of the muscarinic acetylcholine receptor subtype 5 (mAChR5 or M5) with an EC50 of 1.16 μM. It is supplied as a 10 mM solution in DMSO or as a solid and is intended for research use.
  • Positive allosteric modulation of M5 receptor (EC50 1.16 μM).
  • Greater than 30-fold selectivity over M1 and M3 receptors.
  • No potentiator activity observed at M2 or M4 receptors.
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High reported purity (≈99.9%).
Catalog No. 50-002-31931