Learn More
Abcam VU 0238429, allosteric M5 modulator, 25MG
Supplier:  Abcam AB14130725MG
Encompass_Preferred
MW 351.28 Da, Purity >99%. Selective, positive allosteric M5 modulator (EC50 values are 1.16 and >30 μM at M5 and M1/ M3 receptors, respectively). Shows no activity at M2 and M4 receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6696