Learn More
Selleck Chemical LLC VU 0364770 S2862-10mg
SDP
Supplier:  Selleck Chemical LLC S286210MG
Encompass_Preferred
VU 0364770 is a positive allosteric modulator(PAM) of mGlu4 with EC50 of 1 1 M exhibits insignificant activity at 68 other receptors including other mGlu subtypes
Catalog No. 50-311-1867