Learn More
Medchemexpress LLC Vu 0365114 | 1208222-39-2 | 99.4% | 397.35 g·mol⁻1 | C22H14F3NO3 | 100 MG

Supplier: Medchemexpress LLC HY107651100MG
VU 0365114 is a small-molecule selective positive allosteric modulator of the muscarinic acetylcholine receptor M5 for research use. It potentiates acetylcholine-stimulated responses in cellular assays and has been reported to increase insulin secretion in human β-cells. Reported potency is micromolar with minimal activity at M1-M4.
- Selective mAChR M5 positive allosteric modulator with EC50 ~1.6-2.7 μM.
- Demonstrated increase of acetylcholine-stimulated insulin secretion in human β-cells.
- High purity approximately 99.4%.
- Solid, light yellow to orange appearance.
- Molecular formula C22H14F3NO3; molecular weight 397.35 g·mol⁻1.
- Powder stable at -20°C for extended storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.