Learn More
Medchemexpress LLC HY-114933 10mg Medchemexpress, VU0119498 CAS:79183-37-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11493310MG
Encompass_Preferred
Medchemexpress, HY-114933 10mg VU0119498 CAS:79183-37-2 VU0119498 is a pan Gq mAChR M1, M3, M5 positive allosteric modulator (PAM), with EC50s of 6.04, 6.38, and 4.08 µM, respectively. VU0119498 has antidiabetic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0875