Learn More
Abcam VU0152100, M4 positive allosteric modulator, 10MG
Supplier:  Abcam AB14151010MG
Encompass_Preferred
MW 341.4 Da, Purity >99%. Potent and selective M4 mAChr positive allosteric modulator (IC50 = 380 nM). Does not possess intrinsic agonist activity. Inhibits behavioural and neurochemical effects of cocaine. Active *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6545