Learn More
TARGETMOL CHEMICALS INC VU0359595 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T217775MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. VU0359595 (ML-270) is a potent and selective pharmacological phospholipase D1 (PLD1) inhibitor with an IC50 of 3.7 nM. VU0359595 is >1700-fold selective for PLD1 over PLD2 (IC50 of 6.4 uM). VU0359595 can be used for the research of cancer diabetes neurodegenerative and inflammatory diseases. purity: 99%
Catalog No. 50-258-1005