Learn More
Medchemexpress LLC VU0364770 hydrochloride | 1414842-70-8 | 99.7% | 269.13 g/mol | C12H10Cl2N2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100588A10MM 1ML
Encompass_Preferred
VU0364770 hydrochloride is a selective and potent positive allosteric modulator of the metabotropic glutamate receptor 4 (mGlu4), supplied as a ready-to-use 10 mM solution in DMSO for laboratory research. The compound has reported physicochemical identifiers and storage recommendations for reliable experimental use.
Catalog No. 50-002-13642