Learn More
Medchemexpress LLC VU0405601 25mg | 712325-30-9 | 357.20 | C17H13BrN2O2 | 25 MG

Supplier: Medchemexpress LLC HY11675925MG
VU0405601 is a potent KV11.1 (hERG) channel activator reported to protect cardiac tissue from dofetilide-induced ventricular tachycardia. It is supplied as a research-grade small molecule for electrophysiology and cardiac safety studies.
- Potent KV11.1 (hERG) channel activator suitable for electrophysiology studies.
- Reported to protect cardiac tissue from dofetilide-induced ventricular tachycardia in experimental models.
- High purity as listed by supplier (99.93%).
- Molecular formula C17H13BrN2O2; molecular weight 357.20 g/mol.
- Store solid at room temperature up to 3 years; in solvent, store at -80°C or -20°C per supplier guidance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.