Learn More
Medchemexpress LLC VU041 | 332943-64-3 | 99.3% | 363.38 | C19H20F3N3O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11860750MG
Encompass_Preferred
VU041 is a research-grade inhibitor of mosquito inward rectifier potassium 1 (Kir1) channels with submicromolar activity against Anopheles gambiae and Aedes aegypti. Supplied as a high-purity solid for laboratory use; for research use only.
  • Inhibits mosquito Kir1 channels with submicromolar potency.
  • High purity (≈99.3%) suitable for biochemical and electrophysiology assays.
  • Available as a solid 50 MG pack for laboratory use.
  • Soluble in DMSO (≈20 mg/mL) with warming and sonication recommended.
  • Stable when stored as powder at -20°C for long-term retention.
  • Intended for research use only; not for clinical or diagnostic applications.
Catalog No. 50-002-26646