Learn More
Medchemexpress LLC VU0424465 10mM | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY11497810MM 1ML
Encompass_Preferred
VU0424465 is a potent and partial PAM (positive allosteric modulator)-agonist for mGlu5 mediated iCa2 mobilization VU0424465 exhibits high affinity at MPEP allosteric binding site with a Ki value of 11 8 nM VU0424465 is also a agonist for pERK1/2 in cortical neurons[1][2]
Catalog No. 50-002-26892