Learn More
Cayman Chemical VU0453379hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 4139210 MG
Encompass_Preferred
An ago-PAM of GLP-1R; induces calcium mobilization in 9-3-H cells expressing human GLP-1R (EC50 = 1.3 µM); enhances GLP-1-, exenatide-, or liraglutide-induced calcium mobilization in the same cells (EC50s = 1.8, 8.4, and 30 µM, respectively); potentiates exenatide-induced insulin secretion in primary mouse pancreatic islets at 30 µM; reverses haloperidol-induced catalepsy in a rat model of Parkinson’s disease motor symptoms at 30 mg/kg
Catalog No. 50-294-5759