Learn More
Cayman Chemical VU0469650hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 329335 MG
Encompass_Preferred
A negative allosteric mGluR1 modulator; inhibits glutamate-induced calcium flux in cells expressing human mGluR1 (IC50 = 99 nM); >100-fold selective for mGluR1 over mGluR2-8 at 10 µM
Catalog No. 50-295-0351