Learn More
Medchemexpress LLC VU6004256 | 1316652-41-1 | 98.7% | 326.37 | C16H14N4O2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11734850MG
Encompass_Preferred
NCC-149 is a selective histone deacetylase 8 (HDAC8) inhibitor for laboratory research, described for use in neural differentiation studies. It is supplied as a solid powder and is also offered as a 10 mM solution in DMSO. Typical purity is approximately 98.7%, and the molecular weight is 326.37 g·mol⁻¹.
  • Selective HDAC8 inhibitor for neural differentiation research.
  • Available as solid powder and 10 mM solution in DMSO.
  • Typical purity approximately 98.7%.
  • Molecular weight 326.37 g·mol⁻¹ and formula C16H14N4O2S.
  • Intended for laboratory research use only.
Catalog No. 50-002-26798