Learn More
Medchemexpress LLC VU6036720 hydrochloride | 3026597-12-3 | 99.7% | 473.39 | 1 ML

Supplier: Medchemexpress LLC HY148304A10MM/1ML
VU6036720 hydrochloride is a potent and specific in vitro inhibitor of Kir4.1/5.1 channels, demonstrating an IC50 value of 0.24 μM. This compound is suitable for various research applications, particularly those related to brain and kidney studies.
- Potent and specific in vitro Kir4.1/5.1 inhibitor
- Inhibits Kir4.1/5.1 channels with an IC50 of 0.24 μM
- Suitable for brain and kidney research
- High purity of 99.65%
- Supplied as 10 mM solution in 1 mL DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.