Learn More
Medchemexpress LLC VUF11207 fumarate | 1785665-61-3 | 99.9% | 586.65 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11031810MM/1ML
Encompass_Preferred
VUF11207 fumarate is a CXCR7 agonist that specifically binds to CXCR7. It reduces CXCL12-mediated osteoclastogenesis and bone resorption by inhibiting ERK phosphorylation. This product is for research use only and not intended for sale to patients.
  • Acts as a CXCR7 agonist
  • Binds specifically to CXCR7
  • Reduces osteoclastogenesis by inhibiting ERK phosphorylation
  • Reduces bone resorption
Catalog No. 50-003-43294