Learn More
Abcam VUF8430 dihydrobromide, H4 receptor agonist, 50MG
Supplier:  Abcam AB14112850MG
Encompass_Preferred
MW 323.05 g/mol. Potent and high affinity H₄ receptor agonist (pKi = 7.5 and pEC₅₀ = 7.3). Additionally shows activity at H₃ receptors (pKi = 6.5).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6873