Learn More
Cayman Chemical VUF8430hydrobromIde 10mg
SDP
Supplier:  Cayman Chemical 2768210 MG
Encompass_Preferred
A histamine H4 receptor agonist (Ki = 31.6 nM); selective for histamine H4 over H1 and H3 receptors in radioligand binding assays (Kis = >1 and 1 µM, respectively) and is less active in isolated guinea pig atria, which endogenously expresses high levels of H2 receptors (pD2 = 3.8); inhibits forskolin-induced, cAMP-mediated increases in β-galactosidase activity (EC50 = 50.1 nM); enhances HCl-induced formation of gastric lesions in rats at 30 mg/kg; reduces mechanical and thermal allodynia in a mouse model of SNI-induced peripheral neuropathy
Catalog No. 50-293-8369