Learn More
Fuel Cell Earth Carbon Black Vulcan XC-72R, 250 grams
Supplier:  Fuel Cell Earth CVXC72R250
Encompass_Preferred
Vulcan XC 72R carbon powder used as support in the anode and cathode electrodes of Polymer Electrolyte Membrane Fuel Cells (PEMFC), Direct Methanol Fuel Cells (DMFC), Alkaline Fuel Cells (AFC), Microbial Fuel Cells (MFC), Phosphoric Acid Fuel Cells (PAFC), and many more!Vulcan XC 72R carbon black provides excellent conductivity in a range of applications, delivering high conductivity at relatively low loading levels. Among the attributes for Vulcan XC72R are good chemical and physical cleanliness, good process ability, as well as low sulfur content and ionic contamination. Compared to other conductive grades of carbon black, Vulcan XC72R is typically easier to disperse. In addition to conductivity, Vulcan XC72R will also allow a high level of jetness (blackness) to be obtained in the application.
Catalog No. NC2090584