Learn More
TARGETMOL CHEMICALS INC VV116 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T600165MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VV116 (JT001) is a selective and orally active nucleoside antiviral agent against respiratory syncytial virus (RSV) and SARS-CoV-2 infection. purity: 99%
Catalog No. 50-257-9700