Learn More
Selleck Chemical LLC VVD-130037 E4728-5MG
SDP
Supplier:  Selleck Chemical LLC E47285MG
Encompass_Preferred
VVD-130037(BAY-3605349) is an orally available covalent small molecule activator of Kelch-like ECH associated protein 1 (KEAP1) which provide enhanced cellular degradation of nuclear factor-erythroid 2 p45-related factor 2 (NRF2) resulting in significant inhibition of tumor growth in advanced solid tumors
Catalog No. 50-000-41344