Learn More
Andwin Scientific VWR CAPS/BLE 1000/PK
SDP
Supplier:  Andwin Scientific 60819047
Encompass_Preferred
The Plug-Type caps are compatible with 12 x 75 mm and 13 x 100 mm culture tubes. These caps feature a disposable, polyethylene construction, an uncommon fluted structure, and snug fit for either plastic or glass tubes. This product is sold in a pack of 1000 blue caps.
Catalog No. NC9514138