Learn More
Andwin Scientific VWR STAINING DISH GREEN
SDP
Supplier:  Andwin Scientific 25608904
Encompass_Preferred
The Tissue-Tek Slide Staining Dish is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The product is solvent-resistant and is sold in the color of green.
Catalog No. NC9418049