Learn More
Apexbio Technology LLC VX-11e 896720-20-0 10mg
SDP
Supplier:  Apexbio Technology LLC A393110
Encompass_Preferred
VX-11e (CAS 896720-20-0) is a potent and selective inhibitor of extracellular signal-regulated kinase (ERK) a critical serine/threonine kinase within the Ras/Raf/MEK/ERK signaling cascade This pathway is frequently dysregulated in various human cancers and is implicated in mediating oncogenic proliferation and survival signals By directly inhibiting ERK activity VX-11e enables the investigation of ERK-dependent cellular processes and supports the exploration of targeted therapeutic strategies in cancer research
Catalog No. 50-000-95542