Learn More
Selleck Chemical LLC VX-702 10mg 745833-23-2
SDP
Supplier:  Selleck Chemical LLC S600510MG
Encompass_Preferred
VX-702 is a highly selective inhibitor of p38? MAPK, 14-fold higher potency against the p38? versus p38?. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4310