Learn More
Selleck Chemical LLC VX-745 10mg 209410-46-8
SDP
Supplier:  Selleck Chemical LLC S145810MG
Encompass_Preferred
VX-745 is a potent and selective inhibitor of p38? with IC50 of 10 nM, 22-fold greater selectivity versus p38? and no inhibition to p38?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-838-0