Learn More
Medchemexpress LLC HY-19939S 5mg , VX-984 CAS:1476074-39-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19939S5MG
Encompass_Preferred
HY-19939S 5mg VX-984 CAS: 1476074-39-1 VX-984 (M9831) is an orally active, potent, selective and BBB-penetrated DNA-PK inhibitor. VX-984 efficiently inhibits NHEJ (non-homologous end joining) and increases DSBs (DNA double-strand breaks). VX-984 can be used for glioblastomas (GBM) and non-small cell lung cancer (NSCLC) research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9015