Learn More
TARGETMOL CHEMICALS INC VY-3-135 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T97325MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. VY-3-135 is an orally active, stable and specific acetyl-CoA synthetase 2 (ACSS2) inhibitor with IC50 of 44 nM, showing no inhibitory activity against recombinant human ACSS1 or ACSS3. VY-3-135 can effectively inhibit ACSS2-dependent fatty acid metabolism but has no effect on gene expression in tumors. Purity 99.25%
Catalog No. 50-288-0178