Learn More
Selleck Chemical LLC VY-3-135-E1147-100MG
SDP
Supplier:  Selleck Chemical LLC E1147100MG
Encompass_Preferred
VY-3-135 a potent ACSS2 inhibitor with IC50 of 44 3 85 nM potently inhibits ACSS2 dependent fatty acid metabolism but has no effect on gene expression in tumors
Catalog No. 50-313-4902