Learn More
Medchemexpress LLC W-13 hydrochloride | 88519-57-7 | MFCD00058030 | 99.9% | 349.28 g/mol | C14H18Cl2N2O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10091010MM 1ML
Encompass_Preferred
W-13 hydrochloride is a calmodulin antagonist used in research to probe calmodulin-dependent signaling and has been reported to inhibit growth of tamoxifen-resistant human breast cancer cells. The product is offered as solid powder in multiple sizes and as a ready-to-use 10 mM solution in DMSO for cell-based assays.
  • Calmodulin antagonist activity suitable for signaling studies.
  • Reported inhibition of tamoxifen-resistant breast cancer cell growth.
  • Available as solid powder and 10 mM solution in DMSO for convenience.
  • High purity for reproducible experimental results.
  • Molecular weight 349.28 g/mol; CAS 88519-57-7.
Catalog No. 50-002-12257