Learn More
Medchemexpress LLC W-7 hydrochloride | 61714-27-0 | MFCD00012559 | 100.0% | 377.33 g/mol | C16H22Cl2N2O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10091210MM 1ML
Encompass_Preferred
W-7 hydrochloride is a selective calmodulin antagonist used in biochemical and pharmacological research. It inhibits Ca2+-calmodulin-dependent phosphodiesterase and myosin light chain kinase, can induce apoptosis, and is used in studies of Kv4.3 channel function and arrhythmia mechanisms.
  • Selective calmodulin antagonist suitable for biochemical assays.
  • Inhibits Ca2+-calmodulin-dependent enzymes and myosin light chain kinase.
  • Acts as a Kv4.3 channel blocker for electrophysiology research.
  • Available as 10 mM solution in DMSO (1 mL) or as solid in multiple milligram sizes.
  • High purity (≈99.96%) for reproducible results.
  • Identified by CAS 61714-27-0 and standard chemical identifiers for unambiguous reference.
Catalog No. 50-002-12306