Learn More
Medchemexpress LLC W-7 isomer hydrochloride | 69762-85-2 | 99.7% | 377.33 g·mol⁻¹ | C16H22Cl2N2O2S | 1 ML

Supplier: Medchemexpress LLC HY10091310MM 1ML
W-7 isomer hydrochloride is the isomeric hydrochloride salt of W-7, a selective calmodulin antagonist used in biochemical and cell-based research. It is supplied as a solid and as prepared solutions for immediate use.
- High purity: 99.65%.
- Available as a 10 mM solution in DMSO (1 mL) and in solid quantities.
- Molecular formula C16H22Cl2N2O2S; molecular weight 377.33 g·mol⁻¹.
- CAS number 69762-85-2 for unambiguous identification.
- Appearance white to off-white solid.
- SDS, COA, and analytical data available for quality confirmation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.