Learn More
Medchemexpress LLC W-84 dibromide | 21093-51-6 | MFCD00209827 | 98.0% | 708.52 g/mol | C32H44Br2N4O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10097950MG
Encompass_Preferred
W-84 dibromide is a research-grade small molecule described as a potent allosteric modulator and non-competitive antagonist of M2 muscarinic acetylcholine receptors. It retards [3H]N-methylscopolamine dissociation and can stabilize cholinergic antagonist-receptor complexes, making it useful in receptor pharmacology and toxicology research.
  • Potent allosteric modulator of M2 muscarinic receptors.
  • Non-competitive antagonist that stabilizes cholinergic antagonist-receptor complexes.
  • Suitable for receptor pharmacology and organophosphate protection studies.
  • Provided as a solid research chemical with reported high purity.
  • Molecular weight 708.52 g/mol and CAS 21093-51-6 for identification.
Catalog No. 50-002-13146