Learn More
Medchemexpress LLC W-84 dibromide (HDMPPA) | 21093-51-6 | 98.0% | 708.52 | C32H44Br2N4O4 | 100 MG

Supplier: Medchemexpress LLC HY100979100MG
W-84 dibromide (HDMPPA) is a non-competitive muscarinic acetylcholine receptor antagonist and potent allosteric modulator of M2 cholinoceptors. It stabilizes cholinergic antagonist-receptor complexes, retards [3H]N-methylscopolamine dissociation, and has demonstrated over-additive protection against organophosphate intoxication when combined with atropine.
- Purity 98.04%.
- Supplied as a white to off-white solid.
- Molecular weight 708.52 g/mol.
- Molecular formula C32H44Br2N4O4.
- CAS number 21093-51-6.
- Storage sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.