Learn More
MSC ALPHATEC DISP COVERALL
Supplier:  MSC 98850761
Encompass
Disposable & Chemical Resistant Coveralls; Garment Style: Coveralls ; Size: 5X-Large ; Material: Multi-Layer Non-Woven Barrier Laminate Fabric ; Closure Type: Double Zipper; Elastic ; Number Of Pockets: 0.000 ; Cuff Style: Double Cuffs; Elastic
Catalog No. 50-273-6230