Learn More
Cayman Chemical W-MurIcholIc AcId-d4 500ug
SDP
Supplier:  Cayman Chemical 27037500 UG
Encompass_Preferred
An internal standard for the quantification of ω-MCA by GC- or LC-MS
Catalog No. 50-294-0851