Learn More
Medchemexpress LLC W146 trifluoroacetate | 909725-62-8 | 98.2% | 456.39 g/mol | C18H28F3N2O6P | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101395A10MG
Encompass_Preferred
W146 trifluoroacetate is a selective antagonist of the sphingosine-1-phosphate receptor 1 (S1PR1), provided as the trifluoroacetate salt for research use. It is used to inhibit S1P1-mediated signaling in pharmacology and cell-based studies.
  • Selective S1PR1 antagonist (EC50 398 nM; Ki ≈ 70-80 nM).
  • Supplied as a white to off-white trifluoroacetate solid.
  • High purity (98.2%).
  • Available in milligram pack sizes and as ready-to-use DMSO solutions.
  • Store solid at 4°C; solutions stable at -80°C for up to 6 months.
Catalog No. 50-002-12885